Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006339.1 | NP_001006339.1 | 5 - 6 | 536 | 95 | |
GenBank mRNA | AJ720775.1 | - | 5 - 6 | 536 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |