Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006350.2 | NP_001006350.1 | 7 - 8 | 687 | 75 | |
GenBank mRNA | AJ719425.1 | - | 7 - 8 | 689 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |