Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006377.1 | NP_001006377.1 | 7 - 8 | 881 | 123 | |
XM_015282170.2 | XP_015137656.1 | 7 - 8 | 967 | 123 | ||
XM_025147268.1 | XP_025003036.1 | 7 - 8 | 928 | 123 | ||
GenBank mRNA | AJ719818.1 | - | 7 - 8 | 881 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |