Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM206A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006383.1 | NP_001006383.1 | 2 - 3 | 130 | 66 | |
GenBank mRNA | AJ721017.1 | - | 2 - 3 | 130 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |