Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006394.1 | NP_001006394.1 | 8 - 9 | 1500 | 87 | |
GenBank mRNA | AJ720271.1 | - | 8 - 9 | 1500 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |