Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006400.1 | NP_001006400.1 | 2 - 3 | 171 | 90 | |
XM_015283700.2 | XP_015139186.1 | 2 - 3 | 234 | 90 | ||
GenBank mRNA | AJ720519.1 | - | 2 - 3 | 171 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |