Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284055.2 | XP_015139541.1 | 2 - 3 | 5213 | 151 | |
GenBank mRNA | AJ720890.1 | - | 2 - 3 | 186 | 151 | |
BX930977.2 | - | 2 - 3 | 147 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |