Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006406.1 | NP_001006406.1 | 10 - 11 | 1062 | 131 | |
GenBank mRNA | AJ720570.1 | - | 10 - 11 | 1062 | 131 | |
CR352624.1 | - | 10 - 11 | 1046 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |