Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006424.1 | NP_001006424.1 | 5 - 6 | 870 | 106 | |
GenBank mRNA | AJ719570.1 | - | 5 - 6 | 870 | 106 | |
BX933026.1 | - | 3 - 4 | 181 | 106 | ||
CR353503.1 | - | 4 - 5 | 1221 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |