Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006433.2 | NP_001006433.1 | 8 - 9 | 896 | 80 | |
GenBank mRNA | AJ719664.1 | - | 8 - 9 | 898 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |