Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006437.1 | NP_001006437.1 | 8 - 9 | 921 | 66 | |
GenBank mRNA | AJ720791.1 | - | 8 - 9 | 921 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |