Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006445.1 | NP_001006445.1 | 1 - 2 | 122 | 69 | |
GenBank mRNA | AJ720474.1 | - | 1 - 2 | 122 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |