Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006454.1 | NP_001006454.1 | 4 - 5 | 599 | 129 | |
XM_015285436.2 | XP_015140922.1 | 4 - 5 | 716 | 129 | ||
XM_025149692.1 | XP_025005460.1 | 4 - 5 | 935 | 129 | ||
GenBank mRNA | AJ720325.1 | - | 4 - 5 | 599 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |