Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305422.1 | NP_001292351.1 | 5 - 6 | 838 | 123 | |
XM_015285912.2 | XP_015141398.1 | 7 - 8 | 965 | 123 | ||
XM_015285916.2 | XP_015141402.1 | 6 - 7 | 1423 | 123 | ||
XM_015285918.2 | XP_015141404.1 | 7 - 8 | 3423 | 123 | ||
XM_015285920.2 | XP_015141406.1 | 7 - 8 | 1629 | 123 | ||
XM_025149724.1 | XP_025005492.1 | 6 - 7 | 1198 | 123 | ||
XM_025149725.1 | XP_025005493.1 | 7 - 8 | 905 | 123 | ||
XM_025149726.1 | XP_025005494.1 | 6 - 7 | 1537 | 123 | ||
XM_025149727.1 | XP_025005495.1 | 6 - 7 | 1203 | 123 | ||
XM_025149729.1 | XP_025005497.1 | 7 - 8 | 1836 | 123 | ||
XM_025149730.1 | XP_025005498.1 | 6 - 7 | 1216 | 123 | ||
XM_025149731.1 | XP_025005499.1 | 7 - 8 | 3276 | 123 | ||
XM_025149732.1 | XP_025005500.1 | 6 - 7 | 771 | 123 | ||
GenBank mRNA | AJ720126.1 | - | 6 - 7 | 991 | 123 | |
BU142671.1 | - | 3 - 4 | 281 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |