Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006471.1 | NP_001006471.1 | 4 - 5 | 635 | 108 | |
XM_015286612.2 | XP_015142098.1 | 4 - 5 | 639 | 108 | ||
GenBank mRNA | AJ720279.1 | - | 4 - 5 | 635 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |