Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006481.1 | NP_001006481.1 | 7 - 8 | 916 | 65 | |
GenBank mRNA | AJ719909.1 | - | 7 - 8 | 916 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |