Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204554.1 | NP_989885.1 | 6 - 7 | 913 | 123 | |
GenBank mRNA | AJ438290.1 | - | 6 - 7 | 913 | 123 | |
BX930581.2 | - | 6 - 7 | 751 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |