Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204555.1 | NP_989886.1 | 7 - 8 | 597 | 151 | |
XM_025149477.1 | XP_025005245.1 | 8 - 9 | 798 | 151 | ||
XM_025149478.1 | XP_025005246.1 | 8 - 9 | 838 | 151 | ||
XM_025149480.1 | XP_025005248.1 | 7 - 8 | 1059 | 151 | ||
XM_025149481.1 | XP_025005249.1 | 7 - 8 | 791 | 151 | ||
XM_025149482.1 | XP_025005250.1 | 7 - 8 | 748 | 151 | ||
XM_025149483.1 | XP_025005251.1 | 7 - 8 | 818 | 151 | ||
XM_025149484.1 | XP_025005252.1 | 7 - 8 | 695 | 151 | ||
XM_025149485.1 | XP_025005253.1 | 7 - 8 | 1139 | 151 | ||
XM_025149486.1 | XP_025005254.1 | 7 - 8 | 852 | 151 | ||
XM_025149487.1 | XP_025005255.1 | 7 - 8 | 654 | 151 | ||
GenBank mRNA | AF508737.1 | - | 7 - 8 | 597 | 151 | |
DQ099910.1 | - | 7 - 8 | 593 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |