Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204578.4 | NP_989909.3 | 3 - 4 | 570 | 84 | |
GenBank mRNA | AF351205.1 | - | 3 - 4 | 488 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |