Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204579.1 | NP_989910.1 | 7 - 8 | 1234 | 73 | |
GenBank mRNA | AJ431390.1 | - | 7 - 8 | 1234 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |