Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204663.1 | NP_989994.1 | 3 - 4 | 474 | 88 | |
GenBank mRNA | AF204396.2 | - | 3 - 4 | 474 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |