Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204672.1 | NP_990003.1 | 2 - 3 | 282 | 63 | |
XM_015287957.2 | XP_015143443.1 | 2 - 3 | 1413 | 63 | ||
XM_025151467.1 | XP_025007235.1 | 2 - 3 | 1717 | 63 | ||
XM_025151468.1 | XP_025007236.1 | 2 - 3 | 1635 | 63 | ||
XM_025151469.1 | XP_025007237.1 | 3 - 4 | 349 | 63 | ||
GenBank mRNA | AF253513.1 | - | 2 - 3 | 282 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |