Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171601.1 | NP_001165072.1 | 3 - 4 | 766 | 76 | |
NM_204675.2 | NP_990006.2 | 3 - 4 | 664 | 76 | ||
GenBank mRNA | AB024080.1 | - | 3 - 4 | 664 | 76 | |
CR386781.1 | - | 2 - 3 | 741 | 76 | ||
DQ022307.1 | - | 3 - 4 | 769 | 76 | ||
EF068232.1 | - | 3 - 4 | 594 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |