Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204678.1 | NP_990009.1 | 6 - 7 | 813 | 118 | |
XM_015276811.2 | XP_015132297.1 | 6 - 7 | 837 | 118 | ||
GenBank mRNA | AF282244.1 | - | 6 - 7 | 813 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |