Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204697.2 | NP_990028.1 | 2 - 3 | 647 | 76 | |
GenBank mRNA | AF204170.1 | - | 2 - 3 | 648 | 76 | |
AF208401.1 | - | 2 - 3 | 679 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |