Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204704.1 | NP_990035.1 | 3 - 4 | 318 | 126 | |
XM_025154419.1 | XP_025010187.1 | 4 - 5 | 126 | |||
XM_025154420.1 | XP_025010188.1 | 3 - 4 | 126 | |||
GenBank mRNA | AF098513.1 | - | 3 - 4 | 347 | 126 | |
AJ294705.1 | - | 3 - 4 | 318 | 126 | ||
AJ720254.1 | - | 3 - 4 | 424 | 126 | ||
U79757.1 | - | 2 - 3 | 270 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |