Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204705.1 | NP_990036.1 | 1 - 2 | 435 | 93 | |
XM_015280241.2 | XP_015135727.1 | 1 - 2 | 478 | 93 | ||
GenBank mRNA | AF227683.1 | - | 1 - 2 | 435 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |