Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204810.1 | NP_990141.1 | 2 - 3 | 212 | 86 | |
GenBank mRNA | AF027380.1 | - | 2 - 3 | 211 | 86 | |
AF138280.1 | - | 2 - 3 | 212 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |