Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204862.1 | NP_990193.1 | 6 - 7 | 1113 | 87 | |
XM_015278189.2 | XP_015133675.1 | 6 - 7 | 2351 | 87 | ||
XM_015278190.2 | XP_015133676.1 | 6 - 7 | 2348 | 87 | ||
XM_015278191.2 | XP_015133677.1 | 6 - 7 | 2351 | 87 | ||
GenBank mRNA | AJ238613.1 | - | 6 - 7 | 1113 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |