Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204862.1 | NP_990193.1 | 7 - 8 | 1311 | 106 | |
XM_015278189.2 | XP_015133675.1 | 7 - 8 | 2549 | 106 | ||
XM_015278190.2 | XP_015133676.1 | 7 - 8 | 2546 | 106 | ||
XM_015278191.2 | XP_015133677.1 | 7 - 8 | 2549 | 106 | ||
GenBank mRNA | AJ238613.1 | - | 7 - 8 | 1311 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |