Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204880.2 | NP_990211.1 | 2 - 3 | 517 | 74 | |
GenBank mRNA | AB013138.1 | - | 2 - 3 | 518 | 74 | |
CR353975.1 | - | 1 - 2 | 342 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |