Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204887.1 | NP_990218.1 | 2 - 3 | 395 | 92 | |
XM_015292954.2 | XP_015148440.1 | 2 - 3 | 982 | 92 | ||
GenBank mRNA | AB006014.1 | - | 2 - 3 | 395 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |