Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204907.1 | NP_990238.1 | 2 - 3 | 226 | 65 | |
XM_015292957.2 | XP_015148443.1 | 2 - 3 | 287 | 65 | ||
XM_015292958.1 | XP_015148444.1 | 4 - 5 | 716 | 65 | ||
XM_025154421.1 | XP_025010189.1 | 4 - 5 | 1074 | 65 | ||
XM_025154423.1 | XP_025010191.1 | 4 - 5 | 405 | 65 | ||
GenBank mRNA | AF004942.1 | - | 2 - 3 | 226 | 65 | |
AF035676.1 | - | 2 - 3 | 236 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |