Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF043493.1 | - | 2 - 3 | 238 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |