Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204923.3 | NP_990254.3 | 11 - 12 | 2741 | 137 | |
GenBank mRNA | AF043493.1 | - | 9 - 10 | 1180 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |