Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205225.2 | NP_990556.1 | 2 - 3 | 153 | 60 | |
GenBank mRNA | AF208228.1 | - | 1 - 2 | 131 | 60 | |
X81968.1 | - | 154 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |