Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205244.2 | NP_990575.1 | 5 - 6 | 918 | 130 | |
GenBank mRNA | X72892.1 | - | 923 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |