Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205365.1 | NP_990696.1 | 5 - 6 | 620 | 70 | |
GenBank mRNA | U13676.1 | - | 5 - 6 | 620 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |