Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205403.1 | NP_990734.1 | 1 - 2 | 208 | 64 | |
XM_015301090.2 | XP_015156576.1 | 1 - 2 | 486 | 64 | ||
GenBank mRNA | L24548.1 | - | 1 - 2 | 208 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |