Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205417.1 | NP_990748.1 | 5 - 6 | 312 | 73 | |
GenBank mRNA | U19926.1 | - | 5 - 6 | 312 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |