Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205462.1 | NP_990793.1 | 2 - 3 | 332 | 99 | |
GenBank mRNA | M33331.1 | - | 2 - 3 | 332 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |