Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205483.2 | NP_990814.2 | 2 - 3 | 160 | 69 | |
XM_015295934.1 | XP_015151420.1 | 2 - 3 | 236 | 69 | ||
GenBank mRNA | BU125044.1 | - | 2 - 3 | 161 | 69 | |
BU125343.1 | - | 105 | 69 | |||
M25774.1 | - | 2 - 3 | 143 | 69 | ||
V00449.1 | - | 114 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |