Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205512.1 | NP_990843.1 | 4 - 5 | 491 | 69 | |
GenBank mRNA | AY574974.1 | - | 4 - 5 | 461 | 69 | |
AY574975.1 | - | 4 - 5 | 464 | 69 | ||
AY822036.1 | - | 4 - 5 | 464 | 69 | ||
AY822037.1 | - | 4 - 5 | 464 | 69 | ||
CR389373.1 | - | 4 - 5 | 365 | 69 | ||
EU477524.1 | - | 4 - 5 | 461 | 69 | ||
M59925.1 | - | 4 - 5 | 491 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |