Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205513.1 | NP_990844.1 | 4 - 5 | 417 | 67 | |
GenBank mRNA | EU404189.1 | - | 4 - 5 | 315 | 67 | |
M14230.1 | - | 4 - 5 | 417 | 67 | ||
X03343.1 | - | 415 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |