Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205532.1 | NP_990863.1 | 2 - 3 | 226 | 128 | |
XM_015291651.2 | XP_015147137.1 | 2 - 3 | 307 | 128 | ||
GenBank mRNA | KX059701.1 | - | 1 - 2 | 163 | 128 | |
KX059702.1 | - | 1 - 2 | 163 | 128 | ||
KX059703.1 | - | 1 - 2 | 163 | 128 | ||
M99499.1 | - | 2 - 3 | 226 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |