Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: GABPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005816.1 | NP_001005816.1 | 6 - 7 | 752 | 64 | |
NM_001012622.1 | NP_001012640.1 | 6 - 7 | 760 | 64 | ||
GenBank mRNA | AB188512.1 | - | 6 - 7 | 760 | 64 | |
AB188513.1 | - | 5 - 6 | 661 | 64 | ||
AJ719707.1 | - | 6 - 7 | 752 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |