Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006355.1 | NP_001006355.1 | 2 - 3 | 192 | 128 | |
GenBank mRNA | AJ719773.1 | - | 2 - 3 | 192 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |