Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006360.1 | NP_001006360.1 | 4 - 5 | 716 | 85 | |
XM_025147197.1 | XP_025002965.1 | 5 - 6 | 1014 | 85 | ||
GenBank mRNA | AJ719598.1 | - | 4 - 5 | 716 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |