Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006360.1 | NP_001006360.1 | 5 - 6 | 830 | 132 | |
XM_025147197.1 | XP_025002965.1 | 6 - 7 | 1128 | 132 | ||
GenBank mRNA | AJ719598.1 | - | 5 - 6 | 830 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |