Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006362.2 | NP_001006362.1 | 3 - 4 | 393 | 145 | |
GenBank mRNA | AJ719745.1 | - | 3 - 4 | 394 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |